Learn More
Ankyrin Brain Antibody (S105-13), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259374F
Catalog No. NBP259374F
Only null left
Add to Cart
Description
Description
Ankyrin Brain Monoclonal specifically detects Ankyrin Brain in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Ankyrin Brain
Monoclonal
FITC
PBS with 0.05% Sodium Azide
ANK2
Synthetic peptide amino acids 203-496 of human Ankyrin-B
100 μL
287
Human, Mouse, Rat
Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S105-13
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Ank2, Ankyrin 2, Ankyrin 2 brain, Ankyrin B, Ankyrin brain, Ankyrin nonerythroid, Brain Ankyrin
Mouse
Protein G purified
Primary
Detects 200kDa. No cross reactivity against Ankyrin-G.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.