Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ankyrin G Polyclonal Antibody
Rabbit Polyclonal Antibody
$533.50 - $1763.00
Specifications
Antigen | Ankyrin G |
---|---|
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143498
|
Invitrogen™
PA5143498 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143596
|
Invitrogen™
PA5143596 |
100 μL |
Each for $533.50
|
|
|||||
Description
Ankyrin G Polyclonal Antibody for Western Blot, ICC/IF
Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintenance and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.Specifications
Ankyrin G | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
ANK3 | |
2900054D09Rik; AI314020; ANK3; ANK-3; AnkG; ankyrin 3; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin 3, node of Ranvier (ankyrin G); ankyrin G; ankyrin-3; Ankyrin-G; brain-specific ankyrin-G; LOW QUALITY PROTEIN: ankyrin-3; MRT37 | |
ANK3 | |
Primary | |
Affinity chromatography |
1 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
G5E8K5, O70511, Q12955 | |
11735, 288, 361833 | |
The C-terminal 398 amino acids of human ankyrin 3 expressed in and purified from E. coli. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title