Learn More
Ankyrin repeat domain 36B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Ankyrin repeat domain 36B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | ankyrin repeat domain 36B, ankyrin repeat domain-containing protein 36B, CLL-associated antigen KW-1, FLJ21281, FLJ90089, KIAA1641, melanoma-associated antigen, MGC149864, MGC149865, MGC167812 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human Ankyrin repeat domain 36B (NP_079466). Peptide sequence LLDASSRHCTYLENGMQDSRKKLDQMRSQFQEIQDQLTATIRCTKEMEGD |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.