Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKZF1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061589V
Description
ANKZF1 Polyclonal antibody specifically detects ANKZF1 in Human samples. It is validated for Western BlotSpecifications
ANKZF1 | |
Polyclonal | |
Western Blot | |
ankyrin repeat and zinc finger domain containing 1, ankyrin repeat and zinc finger domain-containing protein 1, FLJ13144, Zinc finger protein 744, ZNF744FLJ10415 | |
The immunogen recognized by this antibody maps to a region between residue 676 and 726 of human ankyrin repeat and zinc finger domain containing 1 using the numbering given in entry NP_060559.2 (GeneID 55139). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55139 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction