Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin 2 Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562414 Supplier Invitrogen™ Supplier No. PA562414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYSLDSCDLG LLSSPCWRLP GVYWQNGLSP GVQSTLEPST AKPTEFSWPG TQKQQEAPVE EVGQAEEPDR Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 30%.

C5orf39 may act as a receptor for annexin II on marrow stromal cells to induce osteoclast formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin 2 Receptor
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANXA2R
Gene Accession No. Q3ZCQ2
Gene Alias annexin A2 receptor; annexin II receptor; Annexin-2 receptor; ANXA2R; AX2R; AXIIR; C5orf39
Gene Symbols ANXA2R
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Annexin 2 Receptor. Recombinant protein control fragment (Product #RP-101523).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.