Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin 7 Monoclonal Antibody (ZA004), Abbomax

Catalog No. 502204359
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4359 Supplier Abbomax Inc Supplier No. 605500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Human cervix carcinoma Cellular location: Membrane.

Annexin 7 Monoclonal antibody specifically detects Annexin 7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen Annexin 7
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZA004
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene ANXA7
Gene Accession No. P20073
Gene Alias AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN
Host Species Mouse
Immunogen A recombinant protein of human Annexin VII
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.