Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Antibody (ANXA1/6452R), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314056JF646
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Annexin A1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Recombinant full-length human Annexin A1 protein (Uniprot: P04083) | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| ANXA1/6452R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction