Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Antibody (CPTC-ANXA1-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308699
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Annexin A1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, Immunoprecipitation : 2-4 ug/mg protein, Immunohistochemistry-Paraffin : 1-2 ug/mL | |
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
Recombinant full-length human Annexin A1 protein | |
100 μg | |
Protein Kinase, Signal Transduction | |
301 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
CPTC-ANXA1-1 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction