Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP118842
Description
Annexin A1 Polyclonal specifically detects Annexin A1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Peptide ELISA.Specifications
| Annexin A1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Dog, Cow and Pig. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| NP_000691.1 | |
| ANXA1 | |
| Peptide with sequence CQAILDETKGDYEK corresponding to C-Terminus according to NP_000691.1. | |
| 0.1 mg | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Human, Mouse, Rat, Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction