Learn More
Invitrogen™ Annexin A1 Monoclonal Antibody (6B7F8)

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human Annexin A1 shares 88% and 89% amino acid (aa) sequence identity with mouse and rat Annexin A1, respectively. Positive Control - WB: human A549 whole cell, human K562 whole cell, human U-87 MG whole cell. IHC: human colorectal adenocarcinoma tissue, human lung squamous cell carcinoma tissue, human placenta tissue. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | Annexin A1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 6B7F8 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | ANXA1 |
| Gene Accession No. | P04083 |
| Gene Alias | ANAX1; Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; ANX1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; Phospholipase A2 inhibitory protein; RP11-71A24.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.