Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Antibody (2F1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280577
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Annexin A1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry : 1:200 - 1:1000, Immunohistochemistry-Paraffin : 1:200 - 1:1000 | |
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2F1 | |
Unconjugated | |
PBS | |
ANXA1 | |
Partial recombinant human Annexin A1 (between residues 100-300) expressed in E. coli [UniProt P04083] | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
301 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction