Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A1 Rabbit anti-Human, Clone: RM424, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$181.90 - $548.00

Specifications

Antigen Annexin A1
Clone RM424
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB119702
SDP
View Documents
Novus Biologicals
NBP30917725UL
25 μg
Each for $181.90
Only null left
Add to Cart
 
NB119701
SDP
View Documents
Novus Biologicals
NBP309177100UL
100 μg
Each for $548.00
Only null left
Add to Cart
 
Description

Description

Annexin A1 Monoclonal specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Annexin A1
Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:200
Monoclonal
Purified
RUO
Human
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
A peptide corresponding to the N-terminus of human Annexin A1
Primary
Store at -20°C.
RM424
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Protein Kinase, Signal Transduction
PBS, 50% Glycerol, 1% BSA
301
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.