Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A1 Rabbit anti-Human, Mouse, Clone: ANXA1/3869R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50

Specifications

Antigen Annexin A1
Clone ANXA1/3869R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122908
SDP
View Documents
Novus Biologicals
NBP30771720UG
20 μg
Each of 1 for $304.50
Only null left
Add to Cart
 
Description

Description

Annexin A1 Monoclonal specifically detects Annexin A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Annexin A1
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Protein Kinase, Signal Transduction
10 mM PBS with 0.05% BSA
301
IgG κ
Protein A or G purified
ANXA1/3869R
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Recombinant full-length human Annexin A1 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.