Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Annexin A1 Recombinant Rabbit Monoclonal Antibody (SAIC-13B-19)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547893
Description
Specificity: Recognises human ANXA1.
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.
Specifications
Annexin A1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P04083 | |
ANXA1 | |
Peptide AAYLQETGKPLDETLK derived from Annexin A1 conjugated to KLH. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
SAIC-13B-19 | |
Unconjugated | |
ANXA1 | |
ANAX1; Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; ANX1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; Phospholipase A2 inhibitory protein; RP11-71A24.1 | |
Rabbit | |
Protein A | |
RUO | |
301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction