Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A10 Antibody (OTI4D8), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271683IR
Description
Annexin A10 Monoclonal specifically detects Annexin A10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Annexin A10 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANXA10 | |
Full length recombinant protein of human Annexin A10 (NP_009124) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI4D8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
annexin 14, annexin A10, annexin-10, annexin-14, ANX14Annexin-10 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
11199.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction