Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A10 Antibody (OTI4D8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202282
Description
Annexin A10 Monoclonal specifically detects Annexin A10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Annexin A10 | |
Monoclonal | |
0.98 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
ANXA10 | |
Full length human recombinant protein of human ANXA10(NP_009124) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4D8 | |
Unconjugated | |
annexin 14, annexin A10, annexin-10, annexin-14, ANX14Annexin-10 | |
Mouse | |
37.3 kDa | |
0.1 mL | |
Signal Transduction | |
11199 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction