Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A11 Antibody (OTI1C6), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270182APC
Description
Annexin A11 Monoclonal antibody specifically detects Annexin A11 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Annexin A11 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
OTI1C6 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
annexin A11, Annexin-11, ANX1156-kD, CAP50, CAP-50 | |
Full length human recombinant protein of human ANXA11 (NP_001148) produced in HEK293T cell. | |
0.1 mL | |
Cancer | |
311 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction