Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A11 Antibody (OTI1C6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270182IR
Description
Annexin A11 Monoclonal specifically detects Annexin A11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Annexin A11 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANXA11 | |
| Full length recombinant protein of human Annexin A11 (NP_001148) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1C6 | |
| Western Blot, Immunohistochemistry | |
| annexin A11, Annexin-11, ANX1156-kD, CAP50, CAP-50 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 311.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction