Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A11 Polyclonal Antibody

Catalog No. PIPA597461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597461 Supplier Invitrogen™ Supplier No. PA597461

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Annexin A11 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANXA11
Gene Accession No. P50995
Gene Alias 56 kDa autoantigen; A830099O17Rik; annexin; annexin A11; Annexin A11 (Annexin XI) (Calcyclin-associated annexin 50) (CAP-50); annexin XI; Annexin-11; ANX11; Anxa11; autoantigen; autoantigen, 56-kD; calcyclin-associated annexin 50; Calcyclin-associated annexin-50; CAP50; CAP-50; OTTHUMP00000019957; OTTHUMP00000019958; RP11-369J21
Gene Symbols ANXA11
Host Species Rabbit
Immunogen Recombinant Human Annexin A11 protein (276-505AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.