Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553835 Supplier Invitrogen™ Supplier No. PA553835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHSQSYTLSE GSQQLPKGDS QPSTVVQPLS HPSRNGEPEA PQPAKASSPQ GFDVDRDAKK LNKACKGMGT N Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 32%.

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The specific function of this gene has not yet been determined; however, it is associated with the plasma membrane of undifferentiated, proliferating endothelial cells and differentiated villus enterocytes. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANXA13
Gene Accession No. P27216
Gene Alias 1810034H17Rik; Annexin A13; annexin XIII; annexin, intestine-specific; annexin-13; ANX13; Anxa13; AV055219; Intestine-specific annexin; ISA
Gene Symbols ANXA13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Annexin A13. Recombinant protein control fragment (Product #RP-92286).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.