Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Annexin A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554027
Description
Immunogen sequence: AIIEILSGRT SDERQQIKQK YKATYGKELE EVLKSELSGN FEKTALALLD RPSEYAARQL QKAMKGLGTD ESVLIEV Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The specific function of this gene has not yet been determined; however, it is associated with the plasma membrane of undifferentiated, proliferating endothelial cells and differentiated villus enterocytes. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Annexin A13 | |
Polyclonal | |
Unconjugated | |
ANXA13 | |
1810034H17Rik; Annexin A13; annexin XIII; annexin, intestine-specific; annexin-13; ANX13; Anxa13; AV055219; Intestine-specific annexin; ISA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
312 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P27216 | |
ANXA13 | |
Recombinant protein corresponding to Human Annexin A13. Recombinant protein control fragment (Product #RP-92284). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction