Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A2 Antibody (10H9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Annexin A2 |
---|---|
Clone | 10H9 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Annexin A2 Monoclonal antibody specifically detects Annexin A2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
Annexin A2 | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
annexin A2, Annexin II, annexin-2, ANX2, ANX2L4LPC2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, Chromobindin-8, LIP2PAP-IV, Lipocortin II, LPC2DP36, p36, Placental anticoagulant protein IV, Protein I | |
A synthesized peptide derived from Human Annexin A2 [UniProt P07355] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10H9 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
302 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title