Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A2 Monoclonal Antibody (M009)
GREENER_CHOICE

Catalog No. PIMA547645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547645 Supplier Invitrogen™ Supplier No. MA547645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone M009 detects a 36 kDa* protein corresponding to the molecular mass of Annexin A2 on SDS-PAGE immunoblots of human A431, MDA-MB-231, MeWo, HUVEC, and PC3 cells. The antibody also works for ELISA, immunocytochemical labeling, and immunoprecipitation applications. In addition, mass spectrometry analysis of immunoprecipitates using AM0091 in human A431 cell lysates confirms that this antibody only detects Annexin A2 protein.

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M009
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene ANXA2
Gene Accession No. P07355
Gene Alias 33-kDa calcimedin; 33-kDa lymphocyte Ca>2+>- binding protein; 33kDa lymphocyte Ca>2+>- binding protein; 36-kDa calelectrin; annexin 2; annexin A2; annexin A2 isoform 2; annexin II; Annexin-2; ANX2; ANX2L4; ANXA2; AW215814; CAL1H; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; HEL-S-270; LIP2; lipocortin II; LPC2; LPC2D; p36; PAP-IV; Placental anticoagulant protein IV; protein I
Gene Symbols ANXA2
Host Species Mouse
Immunogen Clone M009 was generated from a proprietary antigen related to human Annexin A2 in MDA-MB-231 breast cancer cell line.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.