Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A2 Mouse anti-Human, Mouse, Clone: 3H1B11, Proteintech
SDP

Catalog No. 501736207 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6207 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6207 Supplier Proteintech Group Inc Supplier No. 600511IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Annexin A2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 3H1B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ANXA2
Gene Accession No. P07355, P07356
Gene Alias Annexin 2, annexin A2, Annexin II, ANX2, ANX2L4, ANXA2, CAL1H, Calpactin 1 heavy chain, Calpactin I heavy chain, Chromobindin 8, LIP2, Lipocortin II, LPC2, LPC2D, P36, PAP IV, Protein I
Gene Symbols Anxa2
Host Species Mouse
Immunogen Annexin A2 Fusion Protein Ag1777
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12306, 302
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.