Learn More
Invitrogen™ Annexin A3 Polyclonal Antibody

Description
Immunogen sequence: ISQAYYTVYK KSLGDDISSE TSGDFRKALL TLADGRRDES LKVDEHLAKQ DAQILYKAGE NRWGTDEDKF TEILCLRSFP QLKLTFDEYR NISQKDIVDS IKGELSGHFE DLLLAIVNCV RNTPAFLAER LHRALKGIGT D Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 94%.
Specifications
Specifications
| Antigen | Annexin A3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ANXA3 |
| Gene Accession No. | P12429 |
| Gene Alias | 35-alpha calcimedin; Annexin 3; annexin A3; Annexin III; Annexin III (Lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin3; annexin-3; ANX3; ANXA 3; ANXA3; calcimedin 35-alpha; inositol 1,2-cyclic phosphate 2-phosphohydrolase; LC3; Lipocortin III; LRRGT00047; PAP-III; Placental anticoagulant protein III |
| Gene Symbols | ANXA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.