Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A4 Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $448.50
Specifications
Antigen | Annexin A4 |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551045
|
Invitrogen™
MA551045 |
200 μL |
Each for $448.50
|
|
|||||
FEMA551046
|
Invitrogen™
MA551046 |
20 μL |
Each for $195.80
|
|
|||||
Description
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.Specifications
Annexin A4 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
35-beta calcimedin; 36 kDa zymogen granule membrane-associated protein; AI265406; AIV; annexin 4; annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); annexin-4; ANX IV; ANX4; ANXA 4; Anxa4; ANXIV; AW106930; Carbohydrate-binding protein p33/p41; chromobindin-4; endonexin; endonexin I; epididymis secretory protein Li 274; HEL-S-274; Lipocortin IV; P32.5; p33/41 (annexin IV); PAP-II; PIG28; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; unnamed protein product; Xanx-4; ZAP 36/annexin IV; ZAP36; zymogen granule membrane associated protein | |
ANXA4 | |
IgG1 κ | |
Protein A/G | |
ANXA4 |
C2 | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P08132, P09525 | |
100312960, 307 | |
Recombinant Human ANXA4 protein, Ala2-Asp319 (Accession #P09525). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title