Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA592862

Catalog No. PIPA592862


Only null left
Add to Cart

Description

Description

Immunogen sequence: IEILASRTPE EIRRISQTYQ QQYGRSLEDD IRSDTSFMFQ RVLVSLSAGG RDEGNYLDDA LVRQDAQDLY EAGEKKWGTD EVKFLTVLCS RNRNHLLHVF DEYKRISQKD IEQSIKSETS GSFEDALLAI VKCMRNKSAY FAEKLYKSMK GLGTDDNTLI RVMVSRAEID MLDIRAHFKR LYGKSLYSFI KGDTSGDYRK VLLVLCGGDD; Positive Samples: BxPC-3, MCF7, A-549, SW620, HepG2, Mouse liver, Mouse kidney, Mouse lung, Rat kidney.

Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Annexin A4
Polyclonal
Unconjugated
ANXA4
35-beta calcimedin; 36 kDa zymogen granule membrane-associated protein; AI265406; AIV; annexin 4; annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); annexin-4; ANX IV; ANX4; ANXA 4; Anxa4; ANXIV; AW106930; Carbohydrate-binding protein p33/p41; chromobindin-4; endonexin; endonexin I; epididymis secretory protein Li 274; HEL-S-274; Lipocortin IV; P32.5; p33/41 (annexin IV); PAP-II; PIG28; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; unnamed protein product; Xanx-4; ZAP 36/annexin IV; ZAP36; zymogen granule membrane associated protein
Rabbit
Affinity Chromatography
RUO
11746, 307, 79124
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.56 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09525, P55260, P97429
ANXA4
Recombinant fusion protein containing a sequence corresponding to amino acids 112-321 of human Annexin A4/Annexin IV (NP_0011441).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.