Learn More
Invitrogen™ Annexin A4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578781
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human A549 whole cell, human THP-1 whole cell, human Hacat whole cell, rat stomach tissue, rat lung tissue, mouse stomach tissue, mouse lung tissue. IHC: human breast cancer tissue, human colonic adenom tissue, human liver cancer tissue, human placenta tissue, mouse kidney tissue, rat kidney tissue. Flow: HEL cell.
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
Specifications
Annexin A4 | |
Polyclonal | |
Unconjugated | |
ANXA4 | |
35-beta calcimedin; 36 kDa zymogen granule membrane-associated protein; AI265406; AIV; annexin 4; annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); annexin-4; ANX IV; ANX4; ANXA 4; Anxa4; ANXIV; AW106930; Carbohydrate-binding protein p33/p41; chromobindin-4; endonexin; endonexin I; epididymis secretory protein Li 274; HEL-S-274; Lipocortin IV; P32.5; p33/41 (annexin IV); PAP-II; PIG28; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; unnamed protein product; Xanx-4; ZAP 36/annexin IV; ZAP36; zymogen granule membrane associated protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11746, 307, 79124 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P09525, P55260, P97429 | |
ANXA4 | |
A synthetic peptide corresponding to a sequence in the middle region of human Annexin IV (119-152aa EEIRRISQTYQQQYGRSLEDDIRSDTSFMFQRVL). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.