Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A6 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299419MFV450
Description
Annexin A6 Polyclonal antibody specifically detects Annexin A6 in Mouse samples. It is validated for Western BlotSpecifications
Annexin A6 | |
Polyclonal | |
Western Blot | |
67 kDa calelectrin, annexin A6, Annexin VI, annexin VI (p68), annexin-6, ANX6, calcium-binding protein p68, calelectrin, calphobindin II, calphobindin-II, CBP68, Chromobindin-20, CPB-II, Lipocortin VI, p68, p70, Protein III | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Annexin A6 | |
0.1 mL | |
Endosome Markers | |
309 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction