Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Annexin A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521651
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (94%), Rat (94%), Chicken (85%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima.
Specifications
Annexin A7 | |
Polyclonal | |
Unconjugated | |
ANXA7 | |
AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
310 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.28 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P20073 | |
ANXA7 | |
Recombinant protein encompassing a sequence within the center region of human Annexin VII. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction