Learn More
Invitrogen™ Annexin A7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592049
Description
Immunogen sequence: TYYDAWSLRK AMQGAGTQER VLIEILCTRT NQEIREIVRC YQSEFGRDLE KDIRSDTSGH FERLLVSMCQ GNRDENQSIN HQMAQEDAQR LYQAGEGRLG TDESCFNMIL ATRSFPQLRA TMEAYSRMAN RDLLSSVSRE FSGYVESGLK TILQCALNRP AFFAERLYYA MKGAGTDDST LVRIVVTRSE IDLVQIKQMF AQMYQKTLGT MIAGDTSGDY RRLLLAIVGQ; Positive Samples: SH-SY5Y, NCI-H460, HepG2, SW480, NIH/3T3, Mouse heart, Mouse lung, Rat lung.
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima.
Specifications
Annexin A7 | |
Polyclonal | |
Unconjugated | |
ANXA7 | |
AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11750, 155423, 310 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.04 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20073, Q07076 | |
ANXA7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 237-466 of human ANXA7 (NP_0011471). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.