Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Annexin A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550196
Description
The antibody detects endogenous levels of total ANXA8 protein.
This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
Specifications
Annexin A8 | |
Polyclonal | |
Unconjugated | |
ANXA8 | |
annexin A8; Annexin VIII; annexin-8; ANX8; ANXA8; CH17-360D5.2; VAC-beta; vascular anticoagulant-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
653145 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P13928 | |
ANXA8 | |
Fusion protein corresponding to a region derived from internal residues of human annexin A8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction