Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583865 Supplier Invitrogen™ Supplier No. PA583865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAWWKSWIEQ EGVTVKSSSH FNPDPDAETL YQAMK Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANXA8
Gene Accession No. P13928
Gene Alias annexin A8; Annexin VIII; annexin-8; ANX8; ANXA8; CH17-360D5.2; VAC-beta; vascular anticoagulant-beta
Gene Symbols ANXA8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANXA8. Recombinant protein control fragment (Product #RP-104835).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.