Learn More
Invitrogen™ Annexin A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595411
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human A549 whole cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
Specifications
Annexin A8 | |
Polyclonal | |
Unconjugated | |
ANXA8 | |
annexin A8; Annexin VIII; annexin-8; ANX8; ANXA8; CH17-360D5.2; VAC-beta; vascular anticoagulant-beta | |
Rabbit | |
Affinity chromatography | |
RUO | |
653145 | |
-20°C | |
Lyophilized |
Flow Cytometry, Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P13928 | |
ANXA8 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human Annexin VIII (20-61aa HFNPDPDAETLYKAMKGIGTNEQAIIDVLTKRSNTQRQQIAK). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.