Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550325 Supplier Invitrogen™ Supplier No. PA550325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ANXA9 protein.

The annexins include ANXA9 coded protein, which are a family of calcium-dependent phospholipid-binding proteins. Members of the annexin family contain 4 internal repeat domains, each of which includes a type II calcium-binding site. The calcium-binding sites are required for annexins to aggregate and cooperatively bind anionic phospholipids and extracellular matrix proteins. Annexin A9 is a divergent member of the annexin protein family in which all four homologous type II calcium-binding sites in the conserved tetrad core contain amino acid substitutions that ablate their function. However, structural analysis suggests that the conserved putative ion channel formed by the tetrad core is intact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Gene ANXA9
Gene Accession No. O76027
Gene Alias 1110003P15Rik; 2310069F17Rik; annexin 31; annexin A9; Annexin XXXI; annexin-31; Annexin-9; ANX31; Anxa9; Pemphaxin
Gene Symbols ANXA9
Host Species Rabbit
Immunogen Full length fusion protein of Human ANXA9.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8416
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.