Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin V Antibody (3B9F3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Annexin V |
---|---|
Clone | 3B9F3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
Annexin V Monoclonal specifically detects Annexin V in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Annexin V | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Apoptosis, Apoptosis Detections | |
PBS with 0.05% Sodium Azide | |
308 | |
Purified recombinant fragment of human ANXA5 (AA: 160-320) expressed in E. Coli. | |
Primary | |
36 kDa |
3B9F3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
ANXA5 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title