Learn More
Annexin V/ANXA5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304788AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.Specifications
Annexin V/ANXA5 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P08758 | |
ANXA5 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ANXA5 | |
Annexin A5; Annexin-5; Annexin V; Lipocortin V; Endonexin II; Calphobindin I; CBP-I; Placental anticoagulant protein I; PAP-I; Placental anticoagulant protein 4; PP4; Thromboplastin inhibitor; Vascular anticoagulant-alpha; VAC-alpha; Anchorin CII; PP4; ENX2; ANX5; ANXA5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
308 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.