Learn More
Specifications
Specifications
| Quantity | 100 Tests |
| Synonym | Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha |
| Content And Storage | Store at 4°C in the dark. |
| Product Line | Novus |
| For Use With (Application) | Detection Instrument: Flow Cytometer Assay Time: 40 min |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
