Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Annexin V Apoptosis Kit [Red, 647]
SDP

Catalog No. RD10188090 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10188090 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10188090 Supplier R&D Systems™ Supplier No. NBP350204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Apoptosis Kit

Specifications

Quantity 100 Tests
Synonym Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha
Content And Storage Store at 4°C in the dark.
Product Line Novus
For Use With (Application) Detection Instrument: Flow Cytometer Assay Time: 40 min
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.