Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin V Monoclonal Antibody (M017)
GREENER_CHOICE

Catalog No. PIMA547647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547647 Supplier Invitrogen™ Supplier No. MA547647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone M017 detects a 35 kDa* protein corresponding to the molecular mass of Annexin A5 on SDS-PAGE immunoblots of human A431, MDA-MB-231, MeWo, and PC3 cells, as well as human skin, breast, lung, and brain. The antibody works for ELISA, western blot, and immunoprecipitation. In addition, mass spectrometry analysis of immunoprecipitates using AM0171 in human MDA-MB-231 cell lysates confirms this antibody only detects Annexin A5 protein.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin V
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M017
Concentration ∼0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Anxa5
Gene Accession No. P08758
Gene Alias Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Gene Symbols Anxa5
Host Species Mouse
Immunogen Clone M017 was generated from a proprietary antigen related to human Annexin A5 in the MDA-MB-231 breast cancer cell line.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.