Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin V Mouse anti-Human, FITC, Clone: VAA-33, Novus Biologicals™

Mouse Monoclonal Antibody
$524.00
Specifications
Antigen | Annexin V |
---|---|
Clone | VAA-33 |
Concentration | 0.2mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Description
Annexin V Monoclonal specifically detects Annexin V in Human samples. It is validated for Flow Cytometry.Specifications
Annexin V | |
0.2mg/mL | |
Flow Cytometry | |
FITC | |
Mouse | |
Apoptosis | |
P08758 | |
308 | |
Recombinant Annexin V | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
VAA-33 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
ANXA5 | |
IgG2a | |
Protein G purified | |
NB100-64303 specifically recognizes human Annexin V, a phospholipid binding protein which binds with high affinity and selectivity to phosphatidylserine (PS), in the presence of calcium. The translocation of PS from the cytosolic leaflet to the outer leaflet of the plasma membrane, due to a loss in plasma membrane asymmetry during apoptosis, acts as an in vivo signal for the removal of the dying cell by phagocytosis. Detection of PS using Annexin V therefore provides a reliable diagnostic tool for the indication of early programmed cell death. Annexin V has also been identified as an anticoagulant protein in the blood coagulation cascade, by acting as an inhibitor of prothrombin activation. The presence of antibodies to Annexin V, are associated with systemic lupus erythematosus (SLE), recurrent spontaneous abortions and systemic sclerosis (SSc). Clone VAA-33 has been reported to work in western blotting applications. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title