Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin V Rabbit anti-Human, Mouse, Rat, Clone: 9A5M8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Annexin V |
---|---|
Clone | 9A5M8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Annexin V Monoclonal antibody specifically detects Annexin V in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Annexin V | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Apoptosis Detections | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
308 | |
IgG | |
Affinity purified |
9A5M8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
A synthetic peptide corresponding to a sequence within amino acids 221-320 of human Annexin A5/Annexin V (P08758). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title