Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin VI Polyclonal Antibody, Cyanine3, Bioss
SDP

Catalog No. 502280716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0716 Supplier Bioss Supplier No. BS8997RCY3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Annexin VI Polyclonal antibody specifically detects Annexin VI in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Annexin VI
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine3
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene ANXA6
Gene Accession No. P08133, P14824
Gene Alias 67 kDa calelectrin; Annexin A6; annexin VI; annexin VI (p68); annexin VI, p68; annexin-6; Anx6; ANXA6; AnxVI; AW107198; Cabm; calcium-binding protein 65/67; calcium-binding protein CATA 65/67; calcium-binding protein p68; calelectrin; calphobindin II; calphobindin-II; Camb; CBP 65/67; CBP68; chromobindin-20; CPB-II; lipocortin VI; p68; p70; Protein III; testis secretory sperm-binding protein Li 198a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Annexin VI
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11749, 309
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.