Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO1 Rabbit anti-Human, Unconjugated, Clone: DG1/2564R, Abnova™

Catalog No. 89339587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-587 Supplier Abnova Corporation Supplier No. RAB00608
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

ANO1 Monoclonal antibody specifically detects ANO1 in Human samples. It is validated for IHC-P.

Specifications

Antigen ANO1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DG1/2564R
Conjugate Unconjugated
Description anoctamin 1, calcium activated chloride channel
Gene ANO1
Gene Alias FLJ10261, ORAOV2, TAOS2, TMEM16A
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 2-101 of human TMEM16A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.