Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733011 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3011 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3011 Supplier Proteintech Group Inc Supplier No. 206471AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCCs include the transmembrane proteins TMEM16A and TMEM16B. TMEM16B is predicted to have eight transmembrane domains with both the amino and carboxy termini in the cytoplasm and is expressed in several tissues including olfactory sensory neurons as well as photoreceptors in mammalian retina. Like TMEM16A, TMEM16B is thought to form at least part of CaCCs but has different biophysical characteristics such as voltage dependence and unitary conductance.

Specifications

Antigen ANO2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANO2
Gene Accession No. Q8CFW1, Q9NQ90
Gene Alias ANO2, anoctamin 2, C12orf3, TMEM16B, Transmembrane protein 16B
Gene Symbols ANO2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100361584, 243634, 57101
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.