Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                ANO3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | ANO3 | 
|---|---|
| Concentration | 0.17 mg/mL | 
| Applications | Immunoprecipitation, Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 50-173-2410    | Proteintech Group Inc 194891AP150UL | 150 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $518.60
                                                 |  | |||||
Description
The protein encoded by this gene belongs to the TMEM16 family of predicted membrane proteins, that are also known as anoctamins. While little is known about the function of this gene, mutations in this gene have been associated with some cases of autosomal domit craniocervical dystonia. Cells from individuals with a mutation in this gene exhibited abnormalities in endoplasmic reticulum-dependent calcium signaling. Studies in rat show that the rat ortholog of this protein interacts with, and modulates the activity of a sodium-activated potassium channel. Deletion of this gene caused increased pain sensitivity in the rat model system. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| ANO3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| A2AHL1, Q9BYT9 | |
| 228432, 63982 | |
| Peptide | |
| Primary | |
| -20°C | |
| ANO3 | 
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ANO3, anoctamin 3, C11orf25, GENX 3947, TMEM16C, Transmembrane protein 16C | |
| ANO3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
             
                                                 
                                                