Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572157
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572157 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572157 Supplier Invitrogen™ Supplier No. PA572157
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANO4 Polyclonal Antibody for Western Blot

Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANO4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ANO4
Gene Accession No. Q32M45
Gene Alias ANO4; anoctamin 4; anoctamin-4; TMEM16D; transmembrane protein 16D
Gene Symbols ANO4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 188-216 amino acids from the N-terminal region of human ANO4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.