Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANO5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109393 Supplier Invitrogen™ Supplier No. PA5109393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGDPDLLEVL AEEGEKVNKH IDYSFQMSEQ SLSSRETSFL INEETMPAKR FNLFLRRRLM FQKNQQSKDS IFFRDGIRQI DFVLSYVDDV KKDAELKAER RKEFETNLRK TGLELEIEDK RDSEDGRTYF VKIHAPWEVL VTYAEVLGIK MPIKESDIPR PKHTPISYVL GPVRLPLSVK YPHPEYFTAQ FSRHRQELFL IEDQATFFPS SSRNRIVYYI LSRCPFGIED GKKRFGIERL LNSNTYSSAY PLHDGQYWKP SEPPNPTNER YTLHQNWARF SYFYKEQPLD LIKNYYGEKI.

Ano5 does not exhibit calcium-activated chloride channel (CaCC) activity. The Ano5 gene encodes a member of the anoctamin family of transmembrane proteins. The Ano5 protein is likely a calcium activated chloride channel. Mutations in the Ano5 gene have been associated with gnathodiaphyseal dysplasia. Alternatively spliced transcript variants have been described for Ano5. Diseases associated with ANO5 include Gnathodiaphyseal Dysplasia and Miyoshi Muscular Dystrophy 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANO5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ANO5
Gene Accession No. Q75UR0, Q75V66
Gene Alias 9330162L24; Ano5; anoctamin 5; anoctamin-5; GDD1; gnathodiaphyseal dysplasia 1 protein; Gnathodiaphyseal dysplasia 1 protein homolog; integral membrane protein GDD1; LGMD2L; RGD1564265; Tmem16e; transmembrane protein 16E
Gene Symbols ANO5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ANO5 (NP_998764.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203859, 233246, 308637
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.