Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANO6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588322
Description
Immunogen sequence: IPRLVYYWSF SVPPYGDHTS YTMEGYINNT LSIFKVADFK NKSKGNPYSD LGNHTTCRYR DFRYPPGHPQ EYKHNIYYWH V; Positive Samples: A-549, Mouse liver, Rat brain; Cellular Location: Cell membrane, Multi-pass membrane protein.
May act as a calcium-activated chloride channel.
Specifications
ANO6 | |
Polyclonal | |
Unconjugated | |
ANO6 | |
2900059G15Rik; AA407480; Ano6; anoctamin 6; anoctamin-6; AW554778; BDPLT7; F730003B03Rik; LOW QUALITY PROTEIN: anoctamin-6; SCAN channel; SCTS; small-conductance calcium-activated nonselective cation channel; Tmem16f; Transmembrane protein 16F | |
Rabbit | |
Affinity Chromatography | |
RUO | |
105722, 196527, 315272 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.92 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q4KMQ2, Q6P9J9 | |
ANO6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 750-830 of human ANO6 (NP_0010205272). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction