Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANO6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535240
Description
ANO6 Polyclonal Antibody for Western Blot
May act as a calcium-activated chloride channel.
Specifications
ANO6 | |
Polyclonal | |
Unconjugated | |
ANO6 | |
2900059G15Rik; AA407480; Ano6; anoctamin 6; anoctamin-6; AW554778; BDPLT7; F730003B03Rik; LOW QUALITY PROTEIN: anoctamin-6; SCAN channel; SCTS; small-conductance calcium-activated nonselective cation channel; Tmem16f; Transmembrane protein 16F | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
196527 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q4KMQ2 | |
ANO6 | |
KLH conjugated synthetic peptide between 68-96 amino acids from the N-terminal region of human ANO6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction